missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva SpeedBeads™ Magnetic Neutravidin Coated Particles, 1 um, 1% solids, 0.05% Na azide
Capture of Biotinylated Probes. Their cauliflower-like structure gives Neutravidin beads high binding capacity. Neutravidin SpeedBeads combine fast reaction kinetics and low, non-specific binding for increased throughput and precision in immunoassay and molecular biology applications.
£235.00 - £4713.00
Specifications
Concentration | 1% solids (10mg/mL) |
---|---|
Content And Storage | 2°C to 8°C |
For Use With (Application) | Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets |
Includes | 0.05% Na azide |
Diameter (Metric) | 1 μm |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11528692
|
Cytiva
78152104010150 |
5 mL |
£762.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11854143
|
Cytiva
78152104010350 |
100 mL |
£4713.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11864143
|
Cytiva
78152104011150 |
1 mL |
£235.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Neutravidin beads exhibit high affinity when binding to biotinylated ligands such as proteins, nucleic acids and peptides. They respond quickly to a magnetic field to shorten assay times, improve precision and move faster through viscous solutions
- Available with a 3500-4500 pmol/mg biotin-binding range
Specifications
1% solids (10mg/mL) | |
Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets | |
1 μm |
2°C to 8°C | |
0.05% Na azide |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cytiva SpeedBeads™ Magnetic Neutravidin Coated Particles, 1 um, 1% solids, 0.05% Na azide