Learn More
3M™ Speedglas™ Inner Cover Plate
3M™ Speedglas™ Inner Cover Plate is designed to protect the inside surface of the filter from spark and spatter.
Brand:  3M™ 7000000227
17.22 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15819634
  • £23.25 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Replacement spare part to the original protection plate if any pits or scratches occur
  • For maximum safety and comfort, make it a habit to check your equipment regularly
  • Replace worn out or damaged parts
  • Regularly clean with a soft cloth and replace if any pits or scratches occur
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Transparent
Marked 117 x 77
5/Pk.
Inner Shield
Cover Plate
9100X