missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Speedglas™ Inner Cover Plate
3M™ Speedglas™ Inner Cover Plate is designed to protect the inside surface of the filter from spark and spatter.
£21.20 - £23.25
Specifications
Color | Transparent |
---|---|
For Use With (Equipment) | Inner Shield |
Product Type | Cover Plate |
Quantity | 5/Pk. |
Style | 9100X |
Description
- Replacement spare part to the original protection plate if any pits or scratches occur
- For maximum safety and comfort, make it a habit to check your equipment regularly
- Replace worn out or damaged parts
- Regularly clean with a soft cloth and replace if any pits or scratches occur
Specifications
Transparent | |
Cover Plate | |
9100X |
Inner Shield | |
5/Pk. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title