Learn More
3M™ Speedglas™ Inner Cover Plate 9002X
3M™ Speedglas™ Inner Cover Plate 9002X is designed to protect the inside surface of the filter from spark and spatter.
Brand:  3M™ 7100080229
36.43 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 15710098
  • £219.00 / Pack of 5
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The inner cover plate is designed to protect the inside surface of the filter from spark and spatter
  • Replace anytime the plate becomes deeply pitted, scratched or too dirty to clean with soft cloth or lens paper
  • Regularly clean with a soft cloth and replace if any pits or scratches occur
  • Can be used to increase +1 or +2 dark shades of the welding filter
  • Failure to use an inside protection plate with the filter voids the warranty if spark or spatter cause damage to the glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
3M Speedglas Welding Filter
Inner Cover Plate
53 x 103 cm
Increase 2 shades