missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3M™ Speedglas™ Inner Cover Plate 9002X
3M™ Speedglas™ Inner Cover Plate 9002X is designed to protect the inside surface of the filter from spark and spatter.
£21.20 - £204.00
Specifications
Color | Clear |
---|---|
Dimensions (L x W) | 53 x 103 cm |
For Use With (Equipment) | 3M Speedglas Welding Filter |
Product Type | Inner Cover Plate |
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Application) | Price | Quantity & Availability | |||||
15709637
|
3M™
7000000184 |
Protection |
£21.20
Pack of 5 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
15760098
|
3M™
7100090450 |
Increase 1 shade |
£184.00
Pack of 5 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
15710098
|
3M™
7100080229 |
Increase 2 shades |
£204.00
Pack of 5 |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
Description
- The inner cover plate is designed to protect the inside surface of the filter from spark and spatter
- Replace anytime the plate becomes deeply pitted, scratched or too dirty to clean with soft cloth or lens paper
- Regularly clean with a soft cloth and replace if any pits or scratches occur
- Can be used to increase +1 or +2 dark shades of the welding filter
- Failure to use an inside protection plate with the filter voids the warranty if spark or spatter cause damage to the glass
Specifications
Clear | |
53 x 103 cm | |
3M Speedglas Welding Filter | |
Inner Cover Plate |