missing translation for 'onlineSavingsMsg'
Learn More
Spermidine, 99%, Thermo Scientific Chemicals Product Code.: 11439396

Spermidine, 99%, Thermo Scientific Chemicals

Product Code. 11439396
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
100 g

Product Code. 11439396

Brand: Thermo Scientific Alfa Aesar A19096.03

Please to purchase this item. Need a web account? Register with us today!

Binds to the polyamine modulatory site of NMDA Spermidine is used in electroporation during transferring the DNA into the cell under the electrical impulse. It is also used for purification of DNA-binding proteins. It combines with calcium chloride and utilized for the precipitation of DNA onto microprojectiles for bombardment with a gene gun. It plays an important role to reduce the amount of aging in yeast, flies, worms, and human immune cells by inducing autophagy. It serves as a precursor of spermine and essential for both normal and neoplastic tissue growth.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Binds to the polyamine modulatory site of NMDA Spermidine is used in electroporation during transferring the DNA into the cell under the electrical impulse. It is also used for purification of DNA-binding proteins. It combines with calcium chloride and utilized for the precipitation of DNA onto microprojectiles for bombardment with a gene gun. It plays an important role to reduce the amount of aging in yeast, flies, worms, and human immune cells by inducing autophagy. It serves as a precursor of spermine and essential for both normal and neoplastic tissue growth.

Solubility
Miscible with water, ethanol and ether.

Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-20-9
Molecular Formula C7H19N3
Molecular Weight (g/mol) 145.25
MDL Number MFCD00008229
InChI Key ATHGHQPFGPMSJY-UHFFFAOYSA-N
Synonym spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropylShow MoreShow Less
PubChem CID 1102
ChEBI CHEBI:16610
IUPAC Name N'-(3-aminopropyl)butane-1,4-diamine
SMILES NCCCCNCCCN

Specifications

Melting Point 22°C to 25°C
Density 0.925
Boiling Point 128°C to 130°C (14 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.479
Quantity 1 g
UN Number UN2735
Beilstein 1698591
Sensitivity Air sensitive
Merck Index 14,8742
Solubility Information Miscible with water,ethanol and ether.
Formula Weight 145.25
Percent Purity 99%
Chemical Name or Material Spermidine
Show More Show Less
compliance-icons
Product Identifier
  • Spermidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : N-(3-aminopropyl)-1,4-Butanediamine
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 24126

    Expires2025-02-13

    Days

    18

    Hours

    15

    Minutes

    21

    Seconds

    0

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Spermidine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.