Learn More
Spermidine, 99%, Thermo Scientific Chemicals
£32.20 - £814.00
Chemical Identifiers
CAS | 124-20-9 |
---|---|
Molecular Formula | C7H19N3 |
Molecular Weight (g/mol) | 145.25 |
MDL Number | MFCD00008229 |
InChI Key | ATHGHQPFGPMSJY-UHFFFAOYSA-N |
Synonym | spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl |
PubChem CID | 1102 |
ChEBI | CHEBI:16610 |
IUPAC Name | N'-(3-aminopropyl)butane-1,4-diamine |
SMILES | NCCCCNCCCN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439396
|
Thermo Scientific Alfa Aesar
A19096.03 |
1 g |
£32.20
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11449396
|
Thermo Scientific Alfa Aesar
A19096.06 |
5 g |
£91.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11459396
|
Thermo Scientific Alfa Aesar
A19096.14 |
25 g |
£340.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15423187
|
Thermo Scientific Alfa Aesar
A19096.22 |
100 g |
£814.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Binds to the polyamine modulatory site of NMDA Spermidine is used in electroporation during transferring the DNA into the cell under the electrical impulse. It is also used for purification of DNA-binding proteins. It combines with calcium chloride and utilized for the precipitation of DNA onto microprojectiles for bombardment with a gene gun. It plays an important role to reduce the amount of aging in yeast, flies, worms, and human immune cells by inducing autophagy. It serves as a precursor of spermine and essential for both normal and neoplastic tissue growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBinds to the polyamine modulatory site of NMDA Spermidine is used in electroporation during transferring the DNA into the cell under the electrical impulse. It is also used for purification of DNA-binding proteins. It combines with calcium chloride and utilized for the precipitation of DNA onto microprojectiles for bombardment with a gene gun. It plays an important role to reduce the amount of aging in yeast, flies, worms, and human immune cells by inducing autophagy. It serves as a precursor of spermine and essential for both normal and neoplastic tissue growth.
Solubility
Miscible with water, ethanol and ether.
Notes
Hygroscopic. Incompatible with acids, acid chlorides, acid anhydrides and oxidizing agents.
Chemical Identifiers
124-20-9 | |
145.25 | |
ATHGHQPFGPMSJY-UHFFFAOYSA-N | |
1102 | |
N'-(3-aminopropyl)butane-1,4-diamine |
C7H19N3 | |
MFCD00008229 | |
spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl | |
CHEBI:16610 | |
NCCCCNCCCN |
Specifications
124-20-9 | |
0.925 | |
>110°C (230°F) | |
1.479 | |
UN2735 | |
Air sensitive | |
spermidine, 1,5,10-triazadecane, 4-azaoctamethylenediamine, spermidin, 4-azaoctane-1,8-diamine, n1-3-aminopropyl butane-1,4-diamine, 1,4-butanediamine, n-3-aminopropyl, n-3-aminopropyl butane-1,4-diamine, n-3-aminopropyl-1,4-butane-diamine, 1,4-diaminobutane, n-3-aminopropyl | |
ATHGHQPFGPMSJY-UHFFFAOYSA-N | |
N'-(3-aminopropyl)butane-1,4-diamine | |
1102 | |
145.25 | |
Spermidine |
22°C to 25°C | |
128°C to 130°C (14 mmHg) | |
C7H19N3 | |
MFCD00008229 | |
1698591 | |
14,8742 | |
Miscible with water,ethanol and ether. | |
NCCCCNCCCN | |
145.25 | |
CHEBI:16610 | |
99% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 204-689-0
missing translation for 'rtecsNumber' : EJ7000000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
- Spermidine
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : N-(3-aminopropyl)-1,4-Butanediamine
RUO – Research Use Only