Learn More
Cole-Parmer™ Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer
These Vial Holders for HG-600 Geno/Grinder™ 2010 Homogenizer securely hold vials in place while processing samples.
Brand:  Cole-Parmer™ 2663
Code : NEW
Additional Details : Weight : 5.00000kg
 View more versions of this product
Product Code. 15390462
  • £468.00 / Pack of 2
Estimated Shipment: 20-09-2024
to see stock.
Add to basket
Description
Description
  • Safely grind samples in the HG-600 Geno/Grinder™ 2010.
  • Vial holders are suitable for use with the HG-600 Geno/Grinder™ 2010 Homogenizer.
  • Order homogenizer and vials separately.
Specifications
Specifications
Holder
Spex Geno/Grinder
Aluminum
Ideal for High Throughput Cryogenic Grinding and to Preserve Pesticides, RNA, Proteins and Other Temperature Sensitive Samples
24 x 5 mL Polyethylene Vials (2241-PE), each pair holds up to 48 vials