Learn More
Cole-Parmer™ XRF Spec-Caps™ Cups
These XRF Spec-Caps™ Cups aid in the production of pressed powder sample disks for XRF, OES, and other analytical techniques.
£33.95 - £403.00
Product Code | Brand | Diameter (Metric) | Height (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Height (Metric) | Quantity | Price | Quantity & Availability | |||
15360542
|
Cole-Parmer™
3615 |
33 mm | 8 mm | 100 Pk. |
£45.40
Pack of 100 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15300552
|
Cole-Parmer™
3617 |
38 mm | 9 mm | 100 Pk. |
£55.45
Pack of 100 Qty Price Break Available |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15310552
|
Cole-Parmer™
3617M |
38 mm | 9 mm | 1000 Pk. |
£403.00
Pack of 1000 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15390542
|
Cole-Parmer™
3619AM |
31 mm | 8 mm | 1000 Pk. |
£217.00
Pack of 1000 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15380542
|
Cole-Parmer™
3619A |
31 mm | 8 mm | 100 Pk. |
£33.95
Pack of 100 |
Estimated Shipment: 01-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15340542
|
Cole-Parmer™
3619 |
30 mm | 8 mm | 100 Pk. |
£43.60
Pack of 100 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Aluminum cups aid in the production of pressed-powder sample discs for XRF.
- Reinforces sample disks to resist chipping and breaking.
- Makes sample disks easier to remove from the pellet die.
- Minimizes contamination during sample handling.
- Protects the bore of the pellet die from abrasive samples.
- Shallow, thin-walled aluminum cups form the bottom and sides of the finished pellet to help reinforce XRF pellets.
Specifications
XRF Spectrometers | |
Sample Cup |