Learn More
BD Accuri™ Spherotech 6-Peak Validation Beads (FL4)
8-Peak validation bead tests are designed to fluoresce in all channels on most flow cytometers and cell sorters, to measure subtle differences in fluorescence levels.
Brand:  BD 653145
Product Code. 12877880
Description
Description
  • Check fluorescence sensitivity and resolution.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
6-Peak validation beads
Flow cytometry and Cell sorters
8 mL
Validation Beads
8 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BD Accuri™ Spherotech 6-Peak Validation Beads (FL4) > 6-Peak Validation Beads; Size: 8 mL