Learn More
Sphingomyelin, MedChemExpress
MedChemExpress Sphingomyelin is a eukaryotic sphingolipid and one of the major constituents of cell membranes and particularly abundant in the myelin sheath that surrounds neuronal axons. Sphingomyelin plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction. Sphingomyelin metabolism is associated with various central nervous system diseases and Niemann-Pick disease.

POA - £60.50
Specifications
Chemical Name or Material | Sphingomyelin |
---|---|
CAS | 85187-10-6 |
Color | White |
Purity Grade Notes | Research |
Solubility Information | Ethanol : 25 mg/mL (34.15 mM; Need ultrasonic) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16494778
![]() |
Medchem Express
HY-113498 |
10 mg |
£60.50
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16407527
![]() |
Medchem Express
HY-113498 |
5 mg |
POA
5mg |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Sphingomyelin is a eukaryotic sphingolipid and one of the major constituents of cell membranes and particularly abundant in the myelin sheath that surrounds neuronal axons. Sphingomyelin plays an important role in cell processes, the regulation of inflammatory responses, and signal transduction. Sphingomyelin metabolism is associated with various central nervous system diseases and Niemann-Pick disease.Specifications
Sphingomyelin | |
White | |
Ethanol : 25 mg/mL (34.15 mM; Need ultrasonic) | |
732.08 | |
95.0% | |
Solid |
85187-10-6 | |
Research | |
[Sphingomyelin] | |
732.08 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.