Learn More
Heidolph™ Spider with Distillation Sleeves
For use with rotary evaporators
Brand: Heidolph™ 15-300-005-09
This item is not returnable.
View return policy
Description
Spider come with a standard NS 29/32 joint size but are also available with NS 24/29 ground joints.
Specifications
Glass | |
Rotary evaporators | |
20 mL (per tube) |
Spider with Distillation Sleeve | |
6 distillation sleeves |