Learn More
New Pig™ Spill Kits
Brand:  New Pig™ KIT213
Encompass_Preferred
 View more versions of this product
Product Code. 12953615
  • £90.75 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Specifications
White
15 L
Oils, Coolants, Solvents, Water--Universal
19.45 in.
11 lb.
4 gal.
Polyethylene, Polypropylene, Magnesium Aluminosilicate
3 in. dia. x 10 ft. L PIG™ Blue Absorbent Sock, 3 in. dia. x 48 in. L PIG™ Blue Absorbent Sock, (4) 15 in. W x 20 in. L PIG™ Absorbent Mat Pad, (6) 2 in. W x 6 in. L Tamperproof Seal Label, (2) 18 in. W x 30 in. H Polyethylene Disposal Bags, Bucket with Lid, 6.5 gallons, printed
12.69 in.