missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Split / Splitless Liner with Double Taper
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 09201801
Code : 13
Additional Details : Weight : 0.10000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Agilent and Varian GC instruments
Specifications
Deactivated | |
1/Pk. | |
4 mm | |
Agilent Technologies, Varian GCs |
GC Inlet Liner | |
78.5 mm | |
6.3 mm |