missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Restek™ Split/Splitless Liners for Agilent™ 6890 GCs: Low Pressure Drop
Includes wool packing
Brand: Restek™ 20995
This item is not returnable.
View return policy
Description
- I.D. 4mm
Specifications
IP deactivation | |
7.85 cm | |
IP Low Pressure Drop Inlet Liner | |
0.63 cm | |
Borosilicate Glass |
5/Pk. | |
Deactivated Wool | |
0.4 cm | |
Agilent GCs Equipped with Split/Splitless Inlets |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Restek™ Split/Splitless Liners for Agilent™ 6890 GCs: Low Pressure Drop
Spot an opportunity for improvement?Share a Content Correction