missing translation for 'onlineSavingsMsg'
Learn More

Cole Parmer™ Square Polystyrene Weigh Boats, White, 500/Pk

Rounded corners simplify transfer of powdered or granular samples.

Brand:  Cole Parmer™ 01017-15

 View more versions of this product

Product Code. 15466033

  • £73.50 / Pack of 500
Estimated Shipment: 24-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Rounded corners simplify transfer of powdered or granular samples.

  • Rounded corners facilitate sample transfer.
  • Disposable.
  • White translucent color dishes for traditional applications.
  • Translucent, rubberized polystyrene withstands temperatures to 180°F, flexes for easy pouring.
  • Biologically inert; resistant to dilute acids, aqueous solutions, alcohols, bases.
  • For use with balances, microwave moisture analyzers, hundreds of other sample handling applications.
  • Flat bottom resists tipping.
  • Contoured sides permit no-slip handling.
Specifications

Specifications

Flat
White
Polystyrene
Weighing Pan
180°F
8.9 cm
2.5 cm
100 mL
Single-use
Square
500/Pk.
3.5 in.
1 in.
1 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers