missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 9243
Selective LXR inverse agonist
Brand: Tocris Bioscience™ 6138/50
This item is not returnable.
View return policy
Description
SR 9243 is a selective LXR inverse agonist. Exhibits no significant activity at other nuclear receptors at maximally effective concentration. Inhibits the Warburg effect and lipogenesis by down-regulation of LXR-mediated gene expression. Selectively reduces cell viability and induces apoptosis in cancer cell lines in vitro. Suppresses hepatic steatosis and inhibits growth of tumor xenografts in mice.Specifications
White | |
1613028-81-1 | |
626.62g/mol | |
Store at 4°C | |
50 mg |
Inhibitor | |
C31H32BrNO4S2 | |
Soluble to 20mM in DMSO | |
Inhibits IL-1β |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction