Learn More
SRA1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA560770
Description
Immunogen sequence: AEVEMAELYV KPGNKERGWN DPPQFSYGLQ TQAGGPRRSL LTKRVAAPQD GSPRVPASET SPGP Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 80%.
This gene is involved in transcriptional coactivation by steroid receptors. There is currently data suggesting this gene encodes both a non-coding RNA that functions as part of a ribonucleoprotein complex and a protein coding mRNA. Increased expression of both the transcript and the protein is associated with cancer.
Specifications
SRA1 | |
Polyclonal | |
Unconjugated | |
SRA1 | |
AA959952; LOW QUALITY PROTEIN: steroid receptor RNA activator 1; MGC87674; pp7684; RGD:621148}; Sra; sra1; sra1 {ECO:0000312; SRAP; steroid receptor coactivator; steroid receptor RNA activator 1; steroid receptor RNA activator 1 (complexes with NCOA1); steroid receptor RNA activator protein; Straa1; Strra1; Unknown (protein for MGC:128615) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10011 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HD15 | |
SRA1 | |
Recombinant protein corresponding to Human SRA1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only