missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tango™ SSTR2-bla U2OS Cells
Tango™ SSTR2-bla U2OS Cells contain the human Somatostatin Receptor 2 (SSTR2) linked to a TEV protease site.
Brand: Tango™ K1582
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Gene Symbol: SSTR2
- Accession Number: NM_001050.2
- Cytoplasmic Tail: Tail from AVPR2 GPCR
- Population Status: Single Cell Clone Origin
- The Tango™ SSTR2-bla U2OS cells contain the human Somatostatin Receptor 2 (SSTR2) linked to a TEV protease site and a Gal4-VP16 transcription factor stably integrated into the Tango™ GPCR-bla U2OS parental cell line. This parental cell line stably expresses a beta-arrestin/TEV protease fusion protein and the beta-lactamase (bla) reporter gene under the control of a UAS response element.
- The Tango™ SSTR2-bla U2OS cells have been functionally validated for Z' factor and EC50 concentrations of Somatostatin-14. In addition, Tango™ SSTR2-bla U2OS cells have been tested for assay performance under variable conditions. These data are found in the Validation & Assay Performance Summary in the Manuals and Brochures section. DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin C, and have no apparent toxicity or change in cellular signal transduction.
Specifications
Liquid Nitrogen | |
GPCRs | |
2 x 106 Cells |
Fluorescent | |
SSTR2-bla U2OS cells, SSTR2-bla U2OS Assay Protocol, Certificate of Analysis | |
Dividing Cells |