Learn More
Thermo Scientific™ Stained Brucella Suspensions
Rapidly detect and quantify specific antibodies to Brucella abortus and Brucella melitensis in sera with this stained suspension.
£94.04
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Description | Stained Brucella Suspensions |
Includes | Stained Brucella Suspensions |
Quantity | 5 mL |
Product Code | Brand | Description | Detectable Analytes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Detectable Analytes | Price | Quantity & Availability | ||||
11923282
|
Thermo Scientific™
R30855501 |
Stained Brucella Suspensions | Brucella abortus |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Rapidly detect and quantify specific antibodies to Brucella abortus and Brucella melitensis in sera with Thermo Scientific™ Remel™ Stained Brucella Suspensions. Stained Brucella Suspensions are standardized, smooth suspensions of killed bacteria which have been stained to facilitate reading of agglutination tests.
Use Stained Brucella Suspensions for the qualitative detection of Brucella abortus and Brucella melitensis antibodies in sera in the investigation of pyrexia and in standard Widal tests1, 2, 3.
- Identifies major groups of Brucella serotypes
- Provides accurate, reliable results
- Simple, easy-to-use tests
Serological tests are based on the fact that antibodies in serum, produced in response to exposure to bacterial antigens, will agglutinate bacterial suspensions which carry homologous antigens.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.
Specifications
2°C to 8°C | |
Stained Brucella Suspensions | |
Stained Brucella Suspensions | |
5 mL |