Learn More
B Braun™ Hypodermic Needles
Designed for patient comfort in a broad array of sizes. B Braun™ Hypodermic Needles provide a smooth surface with light silicone coating designed to minimize discomfort.
Brand:  B Braun™ 466564/3
 View more versions of this product
Product Code. 10438881
  • £61.55 / Pack of 100
Edge
Explore more special offers
Description
Description
Unique, highly automated grinding and polishing process and low-friction lubrication is designed to achieve unequaled sharpness and ease of penetration. Transparent hub design provides high visibility to detect flashback.
  • Color-coded hubs conform to ISO 6009 for easy identity
  • Wide range of diameters and lengths
  • Gauges from 18G to 27G
  • Ultra-sharp, tri-beveled stainless steel designed to result in minimal pain
Specifications
Specifications
Polypropylene (Hub), Stainless Chromium Nickel Steel
21 G
For minimal pain upon puncture, single use
Needle
Ultra-sharp Long Tri-bevel
Green
0.8 mm
Yes
120 mm
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
B Braun™ Hypodermic Needles > Length/Needle Diameter: 120mm/0.8mm