missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Stainless Steel Economy Scoops

These Economy Scoops are ideal for general industrial use.

Brand:  Sampling Systems™ A304-1500

Additional Details : Weight : 2.88000kg

 View more versions of this product

Product Code. 15869611

  • £34.98 / Each
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Description

Description

  • Manufactured from spot welded 304 stainless steel
  • With an attractive high polish
  • Light duty construction
  • Not suitable for GMP applications as there are crevices
Specifications

Specifications

Economy Scoop
1500 mL
General industrial use
Stainless Steel (AISI 304)
High polish
345 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers