Learn More
Sampling Systems™ Stainless Steel Economy Scoops
These Economy Scoops are ideal for general industrial use.
Brand:  Sampling Systems™ A304-2000
Additional Details : Weight : 5.95820kg
 View more versions of this product
Product Code. 15879611
  • £52.55 / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
  • Manufactured from spot welded 304 stainless steel
  • With an attractive high polish
  • Light duty construction
  • Not suitable for GMP applications as there are crevices
Specifications
Specifications
Economy Scoop
2000 mL
General industrial use
Stainless Steel (AISI 304)
High polish