missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Funnel

Used for a wide range of applications. Bochem™ Stainless Steel Funnel has a conical shape.

Brand:  Bochem™ 8844

Additional Details : Weight : 0.24300kg

 View more versions of this product

Product Code. 10539671

  • £58.00 / Each
Estimated Shipment: 23-08-2024
to see stock.
Add to basket

Description

Description

  • Handle 
  • With air canal
  • Tube conical shape
  • 18/10 Stainless steel
Specifications

Specifications

200mm
Stainless Steel
220 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers