Learn More
Bochem™ Stainless Steel Funnel
Used for a wide range of applications. Bochem™ Stainless Steel Funnel has a conical shape.
£33.00 - £197.00
Product Code | Brand | Diameter (Metric)Top | Height (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric)Top | Height (Metric) | Price | Quantity & Availability | ||||
10748741
|
Bochem™
8841 |
100mm | 125 mm |
£33.00
Each |
In Stock
Log in to see stock. |
||||
10018510
|
Bochem™
8843 |
150mm | 190 mm |
£48.80
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||||
10663551
|
Bochem™
8722 |
200mm | 180 mm |
£197.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
10231641
|
Bochem™
8721 |
150mm | 155 mm |
£179.00
Each |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
10539671
|
Bochem™
8844 |
200mm | 220 mm |
£58.00
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
||||
10549671
|
Bochem™
8845 |
250mm | 260 mm |
£40.65
Each |
Estimated Shipment: 19-08-2024
Log in to see stock. |
||||
Description
- Handle
- With air canal
- Tube conical shape
- 18/10 Stainless steel
Specifications
Stainless Steel |