Learn More
RETSCH Stainless Steel Grinding Balls
Designed for use with Retsch™ mixer mills. Retsch™ Stainless Steel Grinding Balls are ideal for disruption of biological cells as well as for DNA/RNA extraction.
Brand: RETSCH 22.455.0003
135.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- For use with MM 200 and MM 400 Mixer Mills
Specifications
Grinding Balls | |
5 mm | |
Reaction Vials | |
Stainless Steel |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.