Learn More
Hamilton™ Stainless Steel HC-75 (H+)
Brand:  Hamilton™ 79476
Product Code. 10074722
  • £1135.00 / Each
Estimated Shipment: 23-05-2025
to see stock.
Explore more special offers
Add to basket
Specifications
12 meq/column
HC-75 (H+)
4.1 mm
9 μm
4.1 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Hamilton™ Stainless Steel HC-75 (H+) > HC-75 (H+)*