Learn More
Grant Instruments™ Stainless Steel Lid
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
£216.00 - £507.00
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | ||||||
15162625
|
Grant Instruments™
LST26 |
ST18, ST26 tanks |
£330.63
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
15152625
|
Grant Instruments™
LST12 |
ST12 tank |
£216.00
Each |
Estimated Shipment: 16-04-2025
Log in to see stock. |
||||||
15172625
|
Grant Instruments™
LST38 |
ST38 tank |
£507.00
Each |
Estimated Shipment: 22-04-2025
Log in to see stock. |
||||||
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
- Stainless steel construction
- Gabled with plastic handle
- For use with Grant Stirred Water Baths and Heated Circulators
- Prevents heat from escaping
- Reduces sample contamination
- For continuous use with water above 90°C.
Specifications
Stainless Steel |