Learn More
Grant Instruments™ Stainless Steel Lid
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
Brand:  Grant Instruments™ LST26
 View more versions of this product
Product Code. 15162625
  • £330.63 / Each
Estimated Shipment: 23-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Helps to reduce evaporation/heat loss and avoid sample contamination. Grant Instruments™ Stainless Steel Lid for Grant Stirred Water Baths is for continuous continuous use with water above 90°C.
  • Stainless steel construction
  • Gabled with plastic handle
  • For use with Grant Stirred Water Baths and Heated Circulators
  • Prevents heat from escaping
  • Reduces sample contamination
  • For continuous use with water above 90°C.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Length: 375
ST18, ST26 tanks
90°C
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Stainless Steel Lid > For Use With: ST26