missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Stainless Steel Lid for Baths with Pitched Lift-off
Helps to reduce evaporation and heat loss while avoiding sample contamination. For use with heated circulating baths.
Brand: Fisherbrand™
Additional Details : Weight : 0.90000kg
Description
- Recommended when bath is used constantly with water above 90°C
Specifications
12 L Bath | |
Stainless Steel | |
380 x 260 x 190 mm |