missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Stainless Steel Lid for Baths with Pitched Lift-off
Helps to reduce evaporation and heat loss while avoiding sample contamination. For use with heated circulating baths.
Brand: Fisherbrand™
Additional Details : Weight : 1.30000kg
Description
- Recommended when bath is used constantly with water above 90°C
Specifications
18 to 26 L Bath | |
Stainless Steel | |
470 x 300 x 360 mm |