missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Stainless Steel Measuring Scoop

Ideal for scooping out fixed volumes of powder. Sampling Systems™ Stainless Steel Measuring Scoop is constructed of high quality Stainless Steel for durability.

Brand:  Sampling Systems™ 1900A-10

 View more versions of this product

Product Code. 15849301

  • £73.25 / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured in 316L Stainless Steel
  • Accurately measures powders
Specifications

Specifications

Measuring Spoon
22mm Scoop Diameter and 26mm Scoop Depth
193 mm
Stainless Steel (316L)
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers