missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Rectangular Retort Stand Bases

Excellent for help holding onto bench equipment and glassware. Fisherbrand™ Rectangular Retort Stand Bases are manufactured from blue acrylic finished steel.

Brand:  Fisherbrand™ 3107/3

 View more versions of this product

Product Code. 11538102

  • £60.75 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Without rod
  • With 10mm hole
  • Fitted with rubber feet
  • Threaded at the center of one short side
Specifications

Specifications

Stainless Steel
250 mm
Retort stand base
250 x 160 mm
With Rubber Feet
Blue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.