Learn More
Fisherbrand™ Stainless Steel Sharp Ends Forceps
Provides a non-stick surface with PTFE coating to which very few solid or liquid substances will permanently adhere. Nickel Electro™ Stainless Steel Sharp Ends Forceps provide excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized.
Brand:  Fisherbrand™ 7204
 View more versions of this product
Product Code. 11582113
  • £25.15 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
PTFE coated forceps, providing a non-stick surface to which very few solid or liquid substances will permanently adhere.
  • Chemically resistant and non-contaminating
  • Non-protein binding and providing bio-inert surfaces
  • No critical quantities lost due to sticking
  • No build up on surfaces
  • Ideal for use under severe laboratory conditions
  • Excellent non-wetting properties for easy cleaning and can be steam or chemically sterilized
  • With guide pin
Specifications
Specifications
Sharp
PTFE-coated Stainless Steel
160 mm
Each
Forceps
Serrated Jaws