missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Contec™ Stainless Steel Sieve for Polypropylene Buckets
Contec Stainless Steel Sieve is for use with Contec Polypropylene Buckets.
£323.00
Specifications
Capacity (Metric) | For 15 L Buckets |
---|---|
Material | Stainless Steel |
Autoclavable | Autoclavable |
Capacity (English) | For 3.9 gal. Buckets |
For Use With (Equipment) | Compact Bucket System |
Specifications
For 15 L Buckets | |
Autoclavable | |
Compact Bucket System | |
Non-sterile |
Stainless Steel | |
For 3.9 gal. Buckets | |
1 Ea. |