Learn More
Grant Instruments™ Stainless Steel Sloping Lids
Recommended for use above 60°C. Stainless Steel Sloping Lid avoids contamination, reduces evaporation and saves energy.
Brand:  Grant Instruments™ LU36
 View more versions of this product
Product Code. 11446427
  • £272.00 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For use with Sub Aqua Pro Series Unstirred Water Bath
  • Triangular-shaped stainless steel with plastic handle
  • Choose from the following available sizes:
    • 5L (LU6)
    • 12L (LU14)
    • 18 or 26L (LU28)
    • 34L (LU36)
Specifications
Specifications
SAP34, ST38
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ Stainless Steel Sloping Lids > For Use With: SAP34