Learn More
Endecotts™ Stainless Steel Test Sieve
Woven wire test sieves for all types of laboratory sampling and particle size analysis
Brand:  Endecotts™ 200SIW10.0
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 12717997
  • £152.00 / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
  • Precision frame that ensures consistent nestability
  • Precise aperture in accordance with ISO 3310-1, ASTM E11 or other specifications
  • Natural fillet (free flowing of sample)
  • Totally sealed no crevice to lose material
  • Evenly tensioned mesh that ensures accurate analysis
  • Safe edge (big radius makes it comfortable to handle)
  • Serial number that ensures full traceability
Specifications
Specifications
Woven Stainless Steel Wire
10 mm
200 mm
Stainless Steel
ISO 3310-1 / BS410