Learn More
Endecotts™ Stainless Steel Test Sieve
Woven wire test sieves for all types of laboratory sampling and particle size analysis
Brand: Endecotts™ 300SIW.090
Additional Details : Weight : 0.10000kg
Descripción
- Precision frame that ensures consistent nestability
- Precise aperture in accordance with ISO 3310-1, ASTM E11 or other specifications
- Natural fillet (free flowing of sample)
- Totally sealed no crevice to lose material
- Evenly tensioned mesh that ensures accurate analysis
- Safe edge (big radius makes it comfortable to handle)
- Serial number that ensures full traceability
Especificaciones
Woven Stainless Steel Wire | |
90 μm | |
8 in. |
Stainless Steel | |
ISO 3310-1 / BS410 |