missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Stainless Steel Universal Scissors

Temperature-resistant universal scissors with a plastic handle. Bochem™ Stainless Steel Universal Scissors are constructed of resistant and durable stainless steel.

Brand:  Bochem™ 4150

 View more versions of this product

Product Code. 10517892

  • £18.50 / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stainless steel construction.
  • Plastic handle.
  • Chemically and thermally resistant (up to 145°C).
Specifications

Specifications

Universal Scissors
200 mm
Stainless Steel, Plastic
Sharp/Sharp
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers