Learn More
Buerkle™ Worm-Threaded Hose Clips
Manufactured from stainless steel 1.4016.
Brand:  Buerkle™ 8677-2540
15.44 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 16616703
  • £22.20 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With screw thread
  • Screw is zinc-coated
Specifications
Specifications
Worm-threaded Hose Clip
Stainless Steel
10 Pk.
25 to 40 mm
Hoses
25 to 40 mm