Learn More
Kinematica Standard Dispersing Aggregate for PT 2500 E
Uses two rows of teeth, is suitable for a large spectrum of laboratory applications, and is reliable and efficient.
Brand:  Kinematica 11030069
 View more versions of this product
Product Code. 11980439
  • £1625.00 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Selection and applications:
    • All standard dispersing tasks
    • Grinding of animal and human tissue samples in a variety of fluids and volumes
    • Sample preparation for the extraction and dissolution of organic materials
    • Sample preparation for medical diagnostic procedures as well as DNA analysis
    • Extractions of systems / substances from vegetable-derived samples
    • Disruption of animal and human cells
    • Deagglomeration of united cell structures
    • Intensive mixing
Specifications
Specifications
Standard Dispersing Aggregate
100 mL to 2 L
192 mm
1 Ea.
21 m/s
12 mm
PT-DA 20/2EC-E19