Learn More
Thermo Scientific™ Standard Fluid Transfer Assembly Extension Set
Simplify process connectivity and enable greater efficiency for most upstream and downstream process operations.
Brand: Thermo Scientific™ SH31079.01
751.00 GBP valid until 2024-12-20
Use promo code "21644" to get your promotional price.
Description
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard Labtainer BPCs, Productainer BPCs, single-use bioreactors, mixers, and fermentors right on the shop floor to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common shop floor processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
- One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
- Multiple manufacturing sites ensure security in product supply
- Produced under cGMP conditions in ISO 7 clean areas to ensure a high level of quality
- Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Specifications
Approved for storage at ambient conditions (15°C to 30°C). | |
3.2mm (1/8 in) ID, 1.6mm (1/16 in) wall, C-Flex 374, 1m (3.25 ft) |