Learn More
Koettermann™ Standard Hanging Cabinet, 600x630x366mm
Koettermann™ Standard Hanging Cabinet, 600x630x366mm provides suitable storage to accommodate a variety of laboratory needs.
Brand: Koettermann™ 307-00189
Additional Details : Weight : 21.00000kg
Description
- Wall-mounted cabinet
- One door
- One shelf
- Side panels
Specifications
366 x 600 x 630 mm | |
Left-Swinging Door | |
No |
Wall-mounted Cabinet | |
1 | |
1 |