missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Standard Rack for Upright Freezer, 50 mm Box Height, with Boxes
Standard Rack fits into any upright freezer and can be used with all standard cryoboxes.
£113.00 - £202.00
Specifications
Includes | White cardboard boxes, 9 x 9 cell dividers |
---|---|
Holds | Boxes |
Material | Stainless Steel |
For Use With (Equipment) | Upright Freezers |
Height (Metric) | 50 mm (Box) |
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Array | Price | Quantity & Availability | ||||
15220664
|
Tenak™
TE23134B |
Boxes | 5 x 4 |
£149.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15200674
|
Tenak™
TE23152B |
Boxes | 4 x 5 |
£162.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15220674
|
Tenak™
TE23154B |
Boxes | 5 x 5 |
£183.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15240654
|
Tenak™
TE23120B |
Boxes | 3 x 3 |
£113.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15240664
|
Tenak™
TE23136B |
Boxes | 6 x 4 |
£177.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15240674
|
Tenak™
TE23156B |
Boxes | 6 x 5 |
£202.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15260654
|
Tenak™
TE23122B |
Boxes | 4 x 3 |
£124.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15260664
|
Tenak™
TE23138B |
Boxes | 7 x 4 |
£192.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15270654
|
Tenak™
TE23124B |
Boxes | 5 x 3 | N/A | |||||
15280664
|
Tenak™
TE23150B |
Boxes | 3 x 5 |
£142.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
15290654
|
Tenak™
TE23130B |
Boxes | 3 x 4 |
£124.00
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
Description
- Measure the inside height and depth of your freezer when choosing the right rack
- Racks can be used with all freezer brands
- Constructed of stainless steel
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
White cardboard boxes, 9 x 9 cell dividers | |
Stainless Steel | |
50 mm (Box) |
Boxes | |
Upright Freezers |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title