Learn More
Koettermann™ Standard Table, Heavy Loads, Wheels, 1200x750x750mm
Koettermann™ Standard Table, Heavy Loads, Wheels, 1200x750x750mm is a mobile laboratory table with a durable worktop and is designed for heavier than normal weight bearing.
Brand:  Koettermann™ 507-00011
Product Code. 15508565
  • £2205.00 / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Heavy-duty mobile table
  • TopResist worktop
  • Steel shelf
Specifications
Specifications
Mobile Table
Heavy-Duty
TopResist Worktop
Steel
750 mm
750 mm
1
1200 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Koettermann™ Standard Table, Heavy Loads, Wheels, 1200x750x750mm > Includes: TopResist Worktop