Learn More
Ismatec™ Standard Tubing, Tygon E-LFL
Durable, chemically resistant tubing available in a broad range of sizes.
Marke:  Ismatec™ SCE0391
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 15723329
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen »
Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
  • DEHP free.
  • Longest service life of any clear Tygon tubing material.
  • Excellent choice where transparency and good chemical resistance is needed.
  • Broad chemical resistance.
  • Tasteless.
  • Extremely low particulate spallation.
  • Meets USP Class VI and FDA criteria.
  • Non-Aging.
Spezifikation
Spezifikation
FDA 21 CFR 175.300; US Pharmacopoea Class VI
4.8 mm
8.0 mm
−50°C
Acids: Fair, Alkaline solutions: Fair, Solvents: Not rec., Pressure: Good, Vacuum: Good, Viscous media: Excellent Sterile media: Limited
Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C (250°F); tubing will appear milky. Gas sterilization with ethylene oxide. Not recommended for sterilization with radiation.
74°C
Autoclavable
Tygon E-LFL
7.5 m
1 Roll
I.D. 4.8 mm, O.D. 8.0 mm
Transparent
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Produktzertifizierungen
Sonderangebote
Sonderangebote