Learn More
Gibco™ Starter Pak No. 3
Provides a variety of yeast and soy products, ideal for processes where an animal origin-free medium is preferred.
Brand:  Gibco™ 215368
Product Code. 16279831
  • £242.00 / Each
Explore more special offers
Description
Description
  • It provides conveniently packaged, 100-g, non-GMP samples of these peptones that are best suited for human and animal vaccine production and contains:
    • Bacto Yeast Extract and Gibco Yeast Extract—these peptones contain a mixture of peptides, amino acids, carbohydrates, and vitamins to support optimal growth in microbial species, and are ideal for the production of vaccines
    • Bacto TC Yeastolate—this peptone is ideal for CHO-based applications of biotherapeutic monoclonal antibodies and recombinant proteins, as well as vaccine applications
    • Phytone Peptone and Difco Soytone—these soy-based supplements provide a nutritious source of carbohydrates for successful use in microbial fermentation. Blending these soy peptones with yeast peptones has been shown to provide additional benefit to cultures.
    • Bacto Malt Extract—a water-soluble portion of malted barley, this peptone also provides carbohydrates for a variety of microbial fermentation processes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Dehydrated Culture Media
1 Ea.
Powder
Human and animal vaccine productions
Animal origin-free, yeast and soy
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Manuals
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Gibco™ Starter Pak No. 3 > Quantity: 1 each