missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ StartingBlock™ Blocking Buffer

StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays.

Brand:  Thermo Scientific™ 37578

35.99 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 13414299

  • £59.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

StartingBlock Blocking Buffer is a single purified protein for fast blocking of western blots and ELISA assays. It provides broad compatibility with a wide range of antibodies, antibody combinations, and other protein probing and assay systems. StartingBlock Blocking Buffer is available in PBS and TBS, with and without Tween 20.

Features of StartingBlock Blocking Buffer include:
Strip and re-probe without re-blocking—blots stay blocked even after stripping with Restore Stripping Buffer (Cat. No. 21059)
Compatible with many detection systems—western blot, ELISA, and IHC with antibody or avidin/biotin probes (blocker is serum- and biotin-free)
Short blocking times—less than 15 minutes for nitrocellulose or PVDF membranes; almost instantaneous for polystyrene microplate wells

TRUSTED_SUSTAINABILITY
Specifications

Specifications

StartingBlock (PBS) Blocking Buffer, 100 mL
100 mL
Western Blot, ELISA, Immunohistochemistry (IHC)
Proprietary protein formulation in phosphate-buffered saline, pH 7.5
PBS
Upon receipt store at 4°C.
Blocking Buffer
Liquid
StartingBlock™
1X
Single purified protein
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers