Learn More
Gibco™ StemPro™ BM Mesenchymal Stem Cells
StemPro™ BM Mesenchymal Stem Cells
Brand:  Gibco™ A15652
Product Code. 15373816
Description
Show More
Specifications
Specifications
Passage 4
Adult
Adherent Cell Culture
Human
Normal
1 x 106 Cells
1 x 106 cells
1 vial. Remove from foil pouch and store in liquid nitrogen.
Stem Cell (Mesenchymal)
Cryopreserved
Bone Marrow
Serum-Supplemented or Serum-Free Media
StemPro™
Product Suggestions
Product Suggestions
Videos
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number8900-105 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA15652
Lot NumberA15652 8900-104 Certificate TypeCertificate of Analysis Date05/06/2019 Catalog NumberA15652
Lot Number8900-101 Certificate TypeCertificate of Analysis Date17/05/2019 Catalog NumberA15652
Lot Number8900-103 Certificate TypeCertificate of Analysis Date19/07/2017 Catalog NumberA15652
Lot Number8900-102 Certificate TypeCertificate of Analysis Date14/04/2017 Catalog NumberA15652
Special Offers
Special Offers
Product Content Correction
Product Title
Gibco™ StemPro™ BM Mesenchymal Stem Cells > 1 x 106 cells