missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ StemPro™ Human Adipose-Derived Stem Cells

StemPro™ Human Adipose-Derived Stem Cells

Brand:  Gibco™ R7788115

Product Code. 10217523

  • £520.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Human Adipose Derived Stem Cells (ADSCs) are isolated from human lipoaspirate tissue and cryopreserved from primary cultures. Before being cryopreserved the ADSCs are expanded for one passage in MesenPRO RS™ Medium (Cat. no. 12746-012) - a revolutionary new reduced serum (2%) medium that results in reduced ADSC doubling times. Each lot of ADSCs originates from a single donor of human lipoaspirate tissue. Each vial of ADSCs contains cells that can differentiate into multiple mature cell phenotypes in vitro including adipocytes, osteoblasts and chondrocytes. ADSCs have demonstrated very similar phenotypic and functional characteristics to that of bone marrow-derived mesenchymal stem cells. The ADSCs express a flow cytometry cell surface protein profile as follows: positive CD29, CD44, CD73, CD90, CD105, CD166; negative CD14, CD31, CD45, Lin1. For optimal ADSC growth and expansion, MesenPRO RS™ Medium is recommended.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

StemPro™ Human Adipose Derived Stem Cells (one vial containing ∼1 x 10^6 cells; store in liquid nitrogen)
Cryopreserved
Lipoaspirate
1 Vial
Stem Cell (Mesenchymal)
Human
StemPro™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1001008 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog NumberR7788115
Lot Number1001009 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog NumberR7788115
Lot Number1001005 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR7788115
Lot Number1001004 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR7788115
Lot Number1001006 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberR7788115
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ StemPro™ Human Adipose-Derived Stem Cells > 1 vial

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.